Merck
  • A novel encapsulation of N,N-diethyl-3-methylbenzamide (DEET) favorably modifies skin absorption while maintaining effective evaporation rates.
A novel encapsulation of N,N-diethyl-3-methylbenzamide (DEET) favorably modifies skin absorption while maintaining effective evaporation rates.
Journal of controlled release : official journal of the Controlled Release Society (2012-05-02)
Jennifer I Karr, Tycho J Speaker, Gerald B Kasting
ABSTRAKT
N,N-diethyl-3-methylbenzamide (DEET) is popular insect repellent which is considered safe and effective, yet is subject to considerable skin absorption. Skin absorption decreases effective repellency since less DEET is available for evaporation. We have investigated the extent to which DEET skin absorption can be reduced and evaporation sustained through encapsulation. DEET permeation through human skin in vitro was measured for an ethanolic solution standard and for two novel topical controlled-release formulations in which the DEET active material was temporarily sequestered within a permeable, charged-film microcapsule. Evaporation measurements were gathered using Tenax TA cartridges and a sampling pump drawing air over the skin. Three formulations were studied: a previously reported microcapsule formulation (Formulation A); a newly-developed microcapsule formulation (Formulation B); and a non-encapsulated ethanol control solution. Formulation B led to a 30% reduction in DEET permeation versus control. The two microcapsule DEET formulations exhibited 36-40% higher cumulative evaporation from the skin than did the control. The vapor trapping measurements in vitro show that Formulation B provided more than 48h of effective evaporation rate for repellency, while Formulation A provided less than 35h and the ethanol control less than 15h. This establishes a technical advantage for the controlled-release approach.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N-Diethyl-3-methylbenzamide, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
DEET, PESTANAL®, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.