Merck
[Kinetics of changes in the coefficient of transmission of the adipose tissue in vitro as a result of photodynamic action].
Biofizika (2012-05-10)
V A Dubrovskiĭ, I Iu Ianina, V V Tuchin
ABSTRAKT
Changes in the light transmission coefficient of human adipose cells as a result of photodynamic action have been detected and investigated. Based on the statistical processing of digital micrographs of thin tissue slices, the kinetics of processes occurring in the adipose tissue has been quantitatively estimated. It has been suggested that the effects observed are brought about through the lipolysis of adipose cells with the retention of their structure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brilliant Green, certified by the BSC
Sigma-Aldrich
Brilliant Green, Dye content ≥85 %