Merck
[Advance in study of ginkgolic acid contained in Ginkgo biloba preparations].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-05-10)
Pingping Liu, Suhua Pan
ABSTRAKT
Ginkgo biloba has a very high medicinal value. The flavonol glycosides and terpene lactones contained in G. biloba extract (GBE) have such pharmacological effects as antioxidant, anti-platelet aggregation and memory improvement, enhancement of immune function. However, the ginkgolic acid (GA) contained in GBE is proved to be highly allergenic and cytotoxic, even minimal residual could also cause severe adverse effects. To minimize the potential safety hazards of ginkgo leaf preparations, this study focuses on GA's chemical structure, adverse effects, toxicity and genesis mechanism, desorption and attenuation in the hope of providing a new thought for studies on safety of Ginkgo biloba preparations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ginkgolic acid C15:1, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Ginkgolic acid C15:1, primary reference standard
Informacje o cenach i dostępności nie są obecnie dostępne.