Merck
  • Acute effects of methiocarb on oxidative damage and the protective effects of vitamin E and taurine in the liver and kidney of Wistar rats.
Acute effects of methiocarb on oxidative damage and the protective effects of vitamin E and taurine in the liver and kidney of Wistar rats.
Toxicology and industrial health (2012-05-25)
Sibel Ozden, Betul Catalgol, Selda Gezginci-Oktayoglu, Ayse Karatug, Sehnaz Bolkent, Buket Alpertunga
ABSTRAKT
Methiocarb (MC) is a widely used carbamate pesticide in agriculture and health programs. Although the main molecular mechanism of carbamate toxicity involves acetylcholinesterase inhibition, studies have also implicated the induction of oxidative stress. Therefore, the present study was aimed to evaluate the effect of acute MC exposure on lipid peroxidation, antioxidant defense systems, histological changes in Wistar rats and the protective effect of pretreatment with vitamin E and taurine. A total of 48 rats were randomly divided into six groups. Rats in group I were given corn oil, while those in group III were dosed with vitamin E (100 mg/kg body weight (b.w.)) and in group V were dosed with taurine (50 mg/kg b.w.). Rats in group II were administered with MC only (25 mg/kg b.w., 1/4 of median lethal dose (LD(50))), while those in groups IV and VI were pretreated with vitamin E (100 mg/kg b.w.) and taurine (50 mg/kg b.w.) for 20 days, respectively, and then exposed to MC (25 mg/kg b.w.). The rats administered with MC showed significant increase in the levels of malondialdehyde in the liver and kidney as an index of lipid peroxidation. Levels of glutathione and activities of superoxide dismutase, catalase and glutathione peroxidase were significantly increased, while activity of glutathione reductase remained unchanged in both the tissues after MC treatment. Mild degenerative histological changes were observed in liver tissue, while the changes in kidney tissue were more severe then liver after MC treatment. Pretreatment with vitamin E and taurine resulted in a significant decrease in the lipid peroxidation and alleviating effects on antioxidant defense systems in both the tissues, while protective effects on the histological changes were shown only in kidney when compared with liver. In conclusion, the study has demonstrated that the acute MC exposure in Wistar rats caused oxidative damage on liver and kidney, which were partly ameliorated by the pretreatment of vitamin E and taurine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Methiocarb, PESTANAL®, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.