Merck
Periodate salts as pyrotechnic oxidizers: development of barium- and perchlorate-free incendiary formulations.
Angewandte Chemie (International ed. in English) (2012-05-29)
Jared D Moretti, Jesse J Sabatini, Gary Chen
ABSTRAKT
In a flash: pyrotechnic incendiary formulations with good stabilities toward various ignition stimuli have been developed without the need for barium or perchlorate oxidizers. KIO(4) and NaIO(4) were introduced as pyrotechnic oxidizers and exhibited excellent pyrotechnic performance. The periodate salts may garner widespread use in military and civilian fireworks because of their low hygroscopicities and high chemical reactivities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Potassium periodate, 99.8%, ACS reagent
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Periodic acid, ACS reagent, 99%