Merck
Development and optimization of dextromethorphan hydrobromide oral disintegrating tablets: effect of formulation and process variables.
Pharmaceutical development and technology (2012-08-14)
Haitham Fady Mostafa, Mohamed Abbas Ibrahim, Adel Sakr
ABSTRAKT
Orally disintegrating tablets (ODTs), which disintegrate rapidly (<1 min) in the mouth and do not require water for administration, have become a very popular dosage form. The study aims to develop a simple and inexpensive method of manufacturing ODTs of a sparingly water-soluble drug, Dextromethorphan hydrobromide. Two factors, three levels (3(2)) full factorial design was used to optimize the diluent, microcrystalline cellulose (X(1)) and superdisintegrant, croscarmellose sodium (X(2)) concentrations. Disintegration time, hardness and T(50) values for all the formulations varied from 12.5 to 152.6 s, 3.58 to 4.92 kp and 0.8 to 2.8 min, respectively. The results indicated that the selected variables have a strong influence on disintegration time, hardness and T(50) of the ODTs. The manufactured ODTs formula composed of 30% microcrystalline cellulose in combination with 3% croscarmellose sodium was chosen as optimized formula, as it showed the lowest disintegration time (12.5 ± 1.22 s), low T(50) (0.8 min.) and hard tablets (4.92 ± 0.28 kp) amongst other tested ODTs formulations. Hardness of DM ODTs was not affected by changing the type of superdisintegrant and lubricant. The disintegration time was significantly (p < 0.05) increased by using sodium starch glycolate instead of croscarmellose sodium.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dextromethorphan HBr, certified reference material, pharmaceutical secondary standard
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)
USP
Sodium Starch Glycolate Type A, United States Pharmacopeia (USP) Reference Standard