Merck
  • Development and validation of a liquid chromatography-tandem mass spectrometry method for the simultaneous determination of acrivastine and pseudoephedrine in human plasma and its application in pharmacokinetics.
Development and validation of a liquid chromatography-tandem mass spectrometry method for the simultaneous determination of acrivastine and pseudoephedrine in human plasma and its application in pharmacokinetics.
Arzneimittel-Forschung (2012-09-01)
J-C He, E-F Feng, M Liu, H-L Li, M Tian, Q Zhang, L-C Dong, G-L Xu
ABSTRAKT
A specific, sensitive and accurate liquid chromatography-tandem mass spectrometry (LC-MS/MS) method has been developed for the simultaneous determination of acrivastine and pseudoephedrine in human plasma samples. Plasma samples were processed and analyzed on a Phenomenex Luna 3 μ CN 100A column (150 mm×2.0 mm) eluted with the mobile phase consisting of methanol and 0.01 mol/L ammonium acetate water solution containing 0.1% formic acid (45:55, v/v) at a flow rate of 0.2 mL/min. The analytes were detected by positive ion electrospray ionization in multiple reaction monitoring mode. The transitions of m/z 349→278, m/z 166→148 and m/z 256→167 were monitored for acrivastine, pseudoephedrine and diphenhydramine (IS), respectively. The method was specific and sensitive with a lower limit of quantitation (LLOQ) of 1.52 ng/mL for acrivastine and 8.13 ng/mL for pseudoephedrine. The method showed good linearity in the range of 1.52~606.0 0 ng/mL for acrivastine and 8.13~813.12 ng/mL for pseudoephedrine (r≥0.996). The mean recovery were ranged 91.82% ~ 98.46% for acrivastine and 90.77% ~ 92.05% for pseudoephedrine. Validation results, such as accuracy, precision and repeatability were within the required limits. The method was successfully applied in a pharmacokinetic study of the acrivastine and pseudoephedrine hydrochloride compound capsule in humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acrivastine, ≥98% (HPLC)
Sigma-Aldrich
Triprolidine hydrochloride, ≥99%
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%