Merck
Colistin: an update on the antibiotic of the 21st century.
Expert review of anti-infective therapy (2012-10-04)
Silpak Biswas, Jean-Michel Brunel, Jean-Christophe Dubus, Martine Reynaud-Gaubert, Jean-Marc Rolain
ABSTRAKT
The emergence of multidrug-resistant Gram-negative bacteria that cause nosocomial infections is a growing problem worldwide. Colistin was first introduced in 1952 and was used until the early 1980s for the treatment of infections caused by Gram-negative bacilli. In vitro, colistin has demonstrated excellent activity against various Gram-negative rod-shaped bacteria, including multidrug-resistant Pseudomonas aeruginosa, Acinetobacter baumannii and Klebsiella pneumoniae. Recent clinical findings regarding colistin activity, pharmacokinetic properties, clinical uses, emerging resistance, toxicities and combination therapy have been reviewed. Recent approaches to the use of colistin in combination with other antibiotics hold promise for increased antibacterial efficacy. It is probable that colistin will be the 'last-line' therapeutic drug against multidrug-resistant Gram-negative pathogens in the 21st century.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Colistin Sulfate, Ready Made Solution, 10 mg/mL (in H2O)