Merck
  • Rapid and selective determination of free chlorine in aqueous solution using electrophilic addition to styrene by gas chromatography/mass spectrometry.
Rapid and selective determination of free chlorine in aqueous solution using electrophilic addition to styrene by gas chromatography/mass spectrometry.
Talanta (2012-12-04)
Kengo Wakigawa, Akinaga Gohda, Sunao Fukushima, Takeshi Mori, Takuro Niidome, Yoshiki Katayama
ABSTRAKT
We developed a rapid and selective method for determination of free chlorine in aqueous solution by gas chromatography/mass spectrometry for the first time. Free chlorine was converted to styrene chlorohydrin using electrophilic addition to styrene in sodium acetate buffer solution (pH 5). The chlorine derivative obtained was extracted with chloroform, and then analyzed by GC/MS. The calibration curve showed good linearity from 0.2-100 μg/mL (as available chlorine). The detection limit was 0.1 μg/mL, and the intra- and interday accuracy were measured at concentrations of 10, 50, and 75 μg/mL to be -1.3 to 6.9% (intraday) and 3.8-8.0% (interday) as % Bias. The precision was between 1.4 and 4.5% as % RSD. These results indicate that this method is a superior technique for the identification of free chlorine. This method was successfully applied to quantification in commercial samples and in samples of a criminal case.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Styrene, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.