Merck
Beneficial effects of natural phenolics on levodopa methylation and oxidative neurodegeneration.
Brain research (2012-12-05)
Ki Sung Kang, Noriko Yamabe, Yujing Wen, Masayuki Fukui, Bao Ting Zhu
ABSTRAKT
Levodopa (L-DOPA) is widely used for symptomatic management in Parkinson's disease. We recently showed that (-)-epigallocatechin-3-gallate, a tea polyphenol, not only inhibits L-DOPA methylation, but also protects against oxidative hippocampal neurodegeneration. In the present study, we sought to determine several other common dietary phenolics, namely, tea catechins [(+)-catechin and (-)-epicatechin] and a representative flavonoid (quercetin), for their ability to modulate L-DOPA methylation and to protect against oxidative hippocampal injury. A combination of in vitro biochemical assays, cell culture-based mechanistic analyses, and in vivo animal models was used. While both tea catechins and quercetin strongly inhibit human liver catechol-O-methyltransferase (COMT)-mediated O-methylation of L-DOPA in vitro, only (+)-catechin exerts a significant inhibition of L-DOPA methylation in both peripheral compartment and striatum in rats. The stronger in vivo effect of (+)-catechin on L-DOPA methylation compared to the other dietary compounds is due to its better bioavailability in vivo. In addition, (+)-catechin strongly reduces glutamate-induced oxidative cytotoxicity in HT22 mouse hippocampal neurons in vitro through inactivation of the nuclear factor-κB signaling pathway. Administration of (+)-catechin also exerts a strong neuroprotective effect in the kainic acid-induced oxidative hippocampal neurodegeneration model in rats. In conclusion, (+)-catechin is a dietary polyphenolic that may have beneficial effects in L-DOPA-based treatment of Parkinson patients by inhibiting L-DOPA methylation plus reducing oxidative neurodegeneration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Carbidopa, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
S-(−)-Carbidopa, ≥98% (TLC), powder
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Reserpine, crystallized, ≥99.0% (HPLC)