Merck
Pharmacokinetic and pharmacodynamic alterations in the Roux-en-Y gastric bypass recipients.
Annals of surgery (2012-12-12)
Sweta Tandra, Naga Chalasani, David R Jones, Samer Mattar, Stephen D Hall, Raj Vuppalanchi
ABSTRAKT
We conducted a pharmacokinetic (PK) study and a pharmacodynamic (PD) study to assess whether Roux-en-Y gastric bypass (RYGB) surgery is associated with significant changes to PK and PD of oral medications. The effect of RYGB on oral drug disposition is not well understood. An oral cocktail of probe drugs for major drug-metabolizing enzymes (caffeine, tolbutamide, omeprazole, dextromethorphan, and oral and intravenous midazolam) was administered to 18 RYGB recipients and 18 controls. Timed blood and urine samples were obtained for PK analyses. Forty mg of oral furosemide was administered to 13 RYGB recipients and 14 controls, and urine and blood samples were collected for assessing furosemidePK, and urine volume and urine sodium excretion for PD analyses. Compared with controls, the RYGB group had significantly lower time to maximum plasma concentration (tmax) for caffeine (0.58 ± 0.5 vs 2.1 ± 2.2 hours, P < 0.0001), tolbutamide (1.4 ± 1.8 vs 2.1 ± 2.2 hours, P = 0.0001), omeprazole (1.1 ± 1.1 vs 4.4 ± 1.3 hours, P < 0.0001), and oral midazolam (0.5 ± 0.2 vs 0.7 ± 0.4 hours, P < 0.01). However, maximum plasma concentration, half-life, area under the curve, and oral bioavailability were not different. Compared with controls, the RYGB group had brisk natriuresis, with significantly lower tmax for urine sodium (1.3 ± 0.5 vs 3.1 ± 2.3 hours, P < 0.02) and correspondingly lower tmax for furosemide (1.8 ± 0.3 vs 4.2 ± 1.2 hours, P = 0.006). However, 6-hour urine sodium and 6-hour urine volume were not different between the two groups. RYGB recipients have significantly shorter tmax for the studied orally administered medications, but otherwise no other significant changes in PK were reported.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Supelco
Dextromethorphan HBr, certified reference material
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)