Merck
Matrix gla protein and alkaline phosphatase are differently modulated in human dermal fibroblasts from PXE patients and controls.
The Journal of investigative dermatology (2012-12-12)
Federica Boraldi, Giulia Annovi, Cees Vermeer, Leon J Schurgers, Tommaso Trenti, Roberta Tiozzo, Deanna Guerra, Daniela Quaglino
ABSTRAKT
Mineralization of elastic fibers in pseudoxanthoma elasticum (PXE) has been associated with low levels of carboxylated matrix gla protein (MGP), most likely as a consequence of reduced vitamin K (vit K) availability. Unexpectedly, vit K supplementation does not exert beneficial effects on soft connective tissue mineralization in the PXE animal model. To understand the effects of vit K supplementation and in the attempt to interfere with pathways leading to the accumulation of calcium and phosphate within PXE-mineralized soft connective tissues, we have conducted in vitro studies on dermal fibroblasts isolated from control subjects and from PXE patients. Cells were cultured in standard conditions and in calcifying medium (CM) in the presence of vit K1 and K2, or levamisole, an alkaline phosphatase (ALP) inhibitor. Control and PXE fibroblasts were characterized by a similar dose-dependent uptake of both vit K1 and vit K2, thus promoting a significant increase of total protein carboxylation in all cell lines. Nevertheless, MGP carboxylation remained much less in PXE fibroblasts. Interestingly, PXE fibroblasts exhibited a significantly higher ALP activity. Consistently, the mineralization process induced in vitro by a long-term culture in CM appeared unaffected by vit K, whereas it was abolished by levamisole.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-Levamisole hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Levamisol hydrochloride, VETRANAL®, analytical standard
Levamisole hydrochloride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)
Levamisole hydrochloride, European Pharmacopoeia (EP) Reference Standard