Merck
N-Chlorosuccinimide, an efficient reagent for on-resin disulfide formation in solid-phase peptide synthesis.
Organic letters (2013-01-17)
Tobias M Postma, Fernando Albericio
ABSTRAKT
N-Chlorosuccinimide is described as a widely applicable on-resin disulfide-forming reagent. Disulfide bond formation was completed within 15 min in DMF. This strategy was successfully used in the synthesis of oxytocin and a regioselective synthesis of an α-conotoxin. Moreover, disulfide formation with N-chlorosuccinimide was found to be compatible with oxidation-prone methionine and tryptophan.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Informacje o cenach i dostępności nie są obecnie dostępne.