Merck
Tetany with Plasmodium falciparum infection.
The Journal of the Association of Physicians of India (2013-02-15)
P S Singh, Neha Singh
ABSTRAKT
Plasmodium falciparum is a malarial infection with high morbidity and wide spectrum of atypical presentation. Here we report an unusual presentation of malaria as tetany with alteration in calcium,phosphate and magnesium metabolism Hypocalcaemia in malaria can cause prolonged Q-Tc interval which could be arisk factor for quinine cardiotoxicity and sudden death Hence monitoring of serum calcium in severe malarial infection and cautious use of quinine in such patients is very important in management
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Calcium D-gluconate, 98.0-102.0%, meets USP testing specifications (Not intended for use in preparing injectable dosage forms.)
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard