Merck
  • Analysis of deoxynivalenol, zearalenone, T-2, and HT-2 toxins in animal feed by LC/MS/MS--a critical comparison of immunoaffinity column cleanup with no cleanup.
Analysis of deoxynivalenol, zearalenone, T-2, and HT-2 toxins in animal feed by LC/MS/MS--a critical comparison of immunoaffinity column cleanup with no cleanup.
Journal of AOAC International (2013-03-05)
Hamide Z Senyuva, John Gilbert, Gözde Türköz, David Leeman, Carol Donnelly
ABSTRAKT
A comparison has been made of an LC/MS/MS method using direct analysis of acetonitrile extracts of feed and cereal samples and a method using acetonitrile extraction and subsequent immunoaffinity column (IAC) cleanup. Naturally contaminated samples containing one or more of deoxynivalenol, zearalenone, T-2, and HT-2 toxins were analyzed together with test materials containing known toxin levels. LC/MS/MS ion ratios and peak profiles, repeatability, and LOQs were used as the basis for comparing the two approaches. The method without cleanup had poorer performance than the method with IAC cleanup in terms of identification based on ion ratios compared to standards. Without cleanup, there was more evidence of background interference, and monitored ions were invariably seen against a noisy background. Nevertheless, quantification of samples analyzed without cleanup gave reasonable agreement with the levels found in the same samples that had received IAC cleanup. Repeatability was poorer with no cleanup, and LOQ values were higher for HT-2 and T-2 toxins, but there was no evidence of any adverse effects on MS performance with repeated injections of crude extracts. Overall, it was concluded that LC/MS/MS analysis of samples with no cleanup is adequate for screening, but for definitive measurements (e.g., for food regulatory control purposes) IAC cleanup remains essential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
HT-2 Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
Zearalenone solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Sigma-Aldrich
HT-2 Toxin, ≥98% (HPLC)
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material