Merck
Zephgrabetaine: a new betaine-type amaryllidaceae alkaloid from Zephyranthes grandiflora.
Natural product communications (2013-03-22)
Deepali Katoch, Dharmesh Kumar, Upendra Sharma, Neeraj Kumar, Yogendra S Padwad, Brij Lal, Bikram Singh
ABSTRAKT
Zephgrabetaine (1), a new betaine type Amaryllidaceae alkaloid, along with seven known alkaloids, lycorine, galanthine, lycoramine, hamayne, haemanthamine, tortuosine, and ungeremine were isolated from the bulbs of Zephyranthes grandiflora and their structures elucidated by spectroscopic data analysis. The isolated alkaloids were tested for in vitro cytotoxic activities against two cell lines, C-6 (rat glioma cells) and CHO-K1 (Chinese hamster ovary cells). A dose dependent cytotoxic effect was exhibited by all the alkaloids on these two cancer cell lines with prominent activity of lycorine and haemanthamine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)