Merck
Naphthazarin protects against glutamate-induced neuronal death via activation of the Nrf2/ARE pathway.
Biochemical and biophysical research communications (2013-03-30)
Tae Gen Son, Elisa M Kawamoto, Qian-Sheng Yu, Nigel H Greig, Mark P Mattson, Simonetta Camandola
ABSTRAKT
Nuclear factor E2-related factor 2 (Nrf2)/antioxidant response element (ARE) pathway is an important cellular stress response pathway involved in neuroprotection. We previously screened several natural phytochemicals and identified plumbagin as a novel activator of the Nrf2/ARE pathway that can protect neurons against ischemic injury. Here we extended our studies to natural and synthetic derivatives of plumbagin. We found that 5,8-dimethoxy-1,4-naphthoquinone (naphthazarin) is a potent activator of the Nrf2/ARE pathway, up-regulates the expression of Nrf2-driven genes in primary neuronal and glial cultures, and protects neurons against glutamate-induced excitotoxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Plumbagin from Plumbago indica
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthoquinone, technical grade