Merck
  • Study of phenolic content and urease and alpha-amylase inhibitory activities of methanolic extract of Rumex acetosella roots and its sub-fractions in different solvents.
Study of phenolic content and urease and alpha-amylase inhibitory activities of methanolic extract of Rumex acetosella roots and its sub-fractions in different solvents.
Pakistan journal of pharmaceutical sciences (2013-04-30)
Dildar Ahmed, Qaria Mumtaz Mughal, Saba Younas, Muhammad Ikram
ABSTRAKT
The present study aimed to establish relationship between urease and alpha-amylase inhibitory activities on the one hand and on the other between anti-enzymatic activities and total phenolic contents of the methanolic extract of roots of Rumex acetosella and its fractions in various solvents. The methanolic extract and its fractions in chloroform, ethyl acetate, n-butanol and water showed remarkable inhibitory activities against both urease and alpha-amylase, there was a close correspondence between urease and alpha-amylase inhibitory activities of the plant samples. The n-butanol fraction which had the highest total phenolic content (252.19 ± 2.32 µg of Gallic Acid Equivalents/mg of dry mass of the sample) showed prominent activity against both urease and alpha-amylase indicating a possible role of phenolics in inhibiting the activities of these enzymes. The samples displayed enzyme inhibitory activities in a dose dependent manner and their effectiveness was comparable with that of the standards, thiourea (for urease) and acarbose (for alpha-amylase). The samples were manifold more effective against urease than alpha-amylase; 2.8 mg/mL of MeOH extract produced about 81% inhibition in alpha-amylase activity, while only 10 µg/mL of the extract was required to create the same inhibition in urease activity. The IC50 values of methanolic, chloroform, ethyl acetate, n-butanolic, aqueous and standard solutions were 1.29, 1.31, 1.90, 1.38, 0.85 and 1.20 (mg/mL) respectively against alpha-amylase and 0.99, 3.89, 1.76, 0.91, 0.85 and 0.97 (μg/mL) respectively against urease. The total phenolic content in MeOH, hexane, chloroform, ethyl acetate, n-butanol and water fractions was 108.88 ± 2.65, 43.70 ± 1.90, 34.44 ± 2.30, 230.71 ± 1.78, 252.19 ± 2.32 and 94.07 ± 2.25 respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG