Merck
Safety and efficacy of a novel nasal spray for maxillary dental anesthesia.
Journal of dental research (2013-05-22)
S G Ciancio, M C Hutcheson, F Ayoub, E A Pantera, C T Pantera, D A Garlapo, B D Sobieraj, S A Almubarak
ABSTRAKT
This study examines the efficacy, safety, and tolerability of a nasal spray to induce anesthesia of maxillary teeth. Forty-five healthy adults requiring restoration of one maxillary tooth were randomized in a 1:2 ratio to receive (1) an intra-oral lidocaine-epinephrine injection with buffered saline nasal spray bilaterally, or (2) a tetracaine hydrochloride-oxymetazoline hydrochloride nasal spray bilaterally with sham injection. Primary endpoints were use of rescue anesthesia and patient global pain assessment. Secondary outcomes included vital sign changes, soft-tissue anesthesia, and treatment-emergent adverse events. In intent-to-treat analysis, 25 of 30 patients given nasal spray (83.3%) did not require rescue anesthesia. Proportion of anesthesia successes for nasal spray was significantly different from the hypothesized placebo anesthesia success of 30% (one-sided p value < .0001 by exact binomial test). Mean duration of soft-tissue anesthesia did not differ significantly by treatment for 3 of 4 sites assessed. No serious adverse events or systemic effects were observed. Tetracaine hydrochloride-oxymetazoline nasal spray appears to provide adequate and safe anesthesia for the majority of maxillary dental procedures. Based on the results from this Phase 2 study, pivotal trials are warranted to validate these findings in an expanded patient population.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Sigma-Aldrich
Lidocaine, analytical standard
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epinephrine