- [Transcription factor Kaiso does not interact with hydroxymethylated DNA within CTGCNA sequence context].
[Transcription factor Kaiso does not interact with hydroxymethylated DNA within CTGCNA sequence context].
Molekuliarnaia biologiia (2013-07-31)
S V Zhenilo, O S Musharova, E B Pokhorchuk
PMID23888785
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female