Merck
Visualization of von Willebrand factor multimers by immunoenzymatic stain using avidin-biotin peroxidase complex.
Thrombosis and haemostasis (1986-04-30)
M Aihara, Y Sawada, K Ueno, S Morimoto, Y Yoshida, M de Serres, H A Cooper, R H Wagner
ABSTRAKT
A technique for the detection of von Willebrand factor multimers separated by discontinuous SDS agarose electrophoresis has been developed using non-radioactive compounds. The multimeric patterns were visualized by monospecific anti-human vWF:Ag followed by incubation with biotinylated antibody. After addition of avidin-biotin-peroxidase complex, the peroxidase activity was detected by 4-chloro-1-naphthol, giving sharp bands with a clear background. By this method, the differences of vWF:Ag multimers could be easily observed between normal plasma and the plasmas from variant type vWD (IIA, IIB, platelet-type). Large and intermediate multimers were absent in the plasma with vWD type IIA, while only large multimers were absent in the plasma with vWD IIB and platelet-type. The absence of large multimers was also observed in two commercial FVIII preparations having the ratio of vWF/vWF:Ag 0.18 and 0.63. The preparation with the ratio of 0.63 showed the presence of larger intermediate multimers. Electrophoresis in SDS 1.5% agarose gel revealed triplet structure of each small multimer, and a relative increase of the smallest subband was observed in vWD IIA plasma, platelet-type vWD plasma and commercial FVIII preparations. The procedures described are easy and safe to perform and are useful for screening or classifying cases with vWD in general laboratories.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline
Sigma-Aldrich
4-Chloro-1-naphthol, tablet