Merck
[Use of antioxidants to prevent damage during acute ischemia and reperfusion of the kidneys].
Biulleten' eksperimental'noi biologii i meditsiny (1983-09-01)
M V Bilenko, L N Shelenkova, G Ia Dubur, A Kh Velena
ABSTRAKT
Prophylactic injection of the natural antioxidant alpha-tocopherol and synthetic antioxidants ionol, diludin and 6-mercurascan prevented the development of lesions during acute renal ischemia and subsequent reperfusion. Ionol proved more effective on intraperitoneal injection than on intragastric route of administration. It has been demonstrated that antiischemic effect of ionol is related to its ability to exhibit the process of lipid peroxidation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 95%
Informacje o cenach i dostępności nie są obecnie dostępne.