- A simple and rapid dot-immunobinding assay for zein and other prolamins.
A simple and rapid dot-immunobinding assay for zein and other prolamins.
Analytical biochemistry (1983-07-15)
A Esen, J M Conroy, S Z Wang
PMID6625179
ABSTRAKT
A simple method for the immunochemical assay of zein and other prolamins is described. Alcohol-solubilized zein is spotted onto chromatography paper disks or strips, incubated with antiserum (30-60 min), washed with several changes of Tris-buffered saline (10 min), incubated with protein A-peroxidase (30 min), and washed with several changes of Tris-buffered saline (10 min). the binding of zein-specific antibody is visualized by peroxidase-catalyzed color production from 4-chloro-1-naphthol. The method is simple, inexpensive, and rapid (2 h total time) and has potential application to other insoluble antigens.