- Craviten hydrolysis in plasma in vitro.
Craviten hydrolysis in plasma in vitro.
Polish journal of pharmacology and pharmacy (1981-10-01)
E Pogonowska-Wala
PMID7322949
ABSTRAKT
Craviten hydrolysis in vitro was studied in human, rat and canine plasma and erythrocytes. Craviten was degraded during the incubation with plasma and the hydrolysis was inhibited by trimethoxybenzoic acid, a metabolite of Craviten. Erythrocyte enzymes do not participate in the hydrolysis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Informacje o cenach i dostępności nie są obecnie dostępne.