Merck
Enantioselective cyanosilylation of ketones catalyzed by recyclable polymeric and dimeric Mn(III) salen complexes at room temperature.
Chirality (2008-06-19)
Noor-Ul H Khan, Santosh Agrawal, Rukhsana I Kureshy, Sayed H R Abdi, K Jeya Prathap, Raksh V Jasra
ABSTRAKT
Recyclable polymeric 1 and dimeric 2 chiral Mn(III) salen complexes catalyzed enantioselective cyanosilylation of various ketones in the presence of triphenylphosphine oxide as an additive proceeded smoothly at room temperature, providing excellent yields (up to 98%) and enantiomeric excess (up to 86%) of respective cyanohydrin trimethylsilyl ether. For most of the substrates, the Catalyst 1 showed slightly better reactivity and enantioselecitivity than the Catalyst 2 nevertheless both the catalysts were easily recovered and reused four times with the retention of their efficiency.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethylsilyl cyanide, technical, ≥95% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimethylsilyl cyanide, 98%
Informacje o cenach i dostępności nie są obecnie dostępne.