- Four new coumarin derivatives from Artemisia keiskeana.
Four new coumarin derivatives from Artemisia keiskeana.
Journal of natural products (2001-08-25)
J H Kwak, K B Lee, F J Schmitz
PMID11520232
ABSTRAKT
Four new coumarin monoterpene ethers, artekeiskeanols A-D (1-4), and three known coumarins, isofraxidin, fraxidin, and daphnoretin, were isolated from the whole plants of Artemisia keiskeana. All structures were determined from spectral data, and that of artekeiskeanol A (1) was confirmed by synthesis.