Merck
Effect of novel anti-inflammatory ethanolamine derivatives with antioxidant properties on drug metabolising enzymes.
European journal of drug metabolism and pharmacokinetics (2003-09-25)
I Andreadou, E A Rekka, P N Kourounakis
ABSTRAKT
The influence of four ethanolamine derivatives with anti-inflammatory and antioxidant activity on the in vitro aminopyrine N-demethylation was studied. It was found that these compounds inhibit the N-demethylation of aminopyrine. 1-Cyclohexyl-5-(2-hydroxy-ethylamino)-pentan-2-one (compound 4), possessing the highest inhibitory activity and found earlier to be a potent anti-inflammatory agent, is further tested in vivo on zoxazolamine-induced paralysis, after a single administration to rats, and on aminopyrine N-demethylation, rat hepatic total cytochrome P450 and protein (postmitochondrial and microsomal) content, after a prolonged treatment. It was found that the examined compound had no significant influence on the above biotransformations, however, it could decrease the catalytically active hepatic cytochrome P450 content. These results, considered together with some structural and physicochemical properties of the compound, indicate that this compound may act as a CYP2D6 substrate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Amino-5-chlorobenzoxazole, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.