Merck
Natural antibiotic function of a human gastric mucin against Helicobacter pylori infection.
Science (New York, N.Y.) (2004-08-18)
Masatomo Kawakubo, Yuki Ito, Yukie Okimura, Motohiro Kobayashi, Kyoko Sakura, Susumu Kasama, Michiko N Fukuda, Minoru Fukuda, Tsutomu Katsuyama, Jun Nakayama
ABSTRAKT
Helicobacter pylori infects the stomachs of nearly a half the human population, yet most infected individuals remain asymptomatic, which suggests that there is a host defense against this bacterium. Because H. pylori is rarely found in deeper portions of the gastric mucosa, where O-glycans are expressed that have terminal alpha1,4-linked N-acetylglucosamine, we tested whether these O-glycans might affect H. pylori growth. Here, we report that these O-glycans have antimicrobial activity against H. pylori, inhibiting its biosynthesis of cholesteryl-alpha-D-glucopyranoside, a major cell wall component. Thus, the unique O-glycans in gastric mucin appeared to function as a natural antibiotic, protecting the host from H. pylori infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesterol β-D-glucoside, ≥97% (TLC)
Informacje o cenach i dostępności nie są obecnie dostępne.