Merck
Meniere's disease: an evidence based approach to assessment and management.
International journal of clinical practice (2012-01-20)
I Syed, C Aldren
ABSTRAKT
Menière's disease (MD) is frequently over-diagnosed in both primary and secondary care. This is unfortunate given the significant medical and social implications of such a diagnosis. Difficulties may arise in differentiating the patient with true MD from those individuals with less clearly defined disorders of cochleo-vestibular function. In this review, we suggest a practical evidence based approach to assessment and management of the patient with MD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-(2-Methylaminoethyl)pyridine, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.