Merck
Magneto-controlled electrochemical immunoassay of brevetoxin B in seafood based on guanine-functionalized graphene nanoribbons.
Biosensors & bioelectronics (2012-06-12)
Juan Tang, Li Hou, Dianping Tang, Jun Zhou, Zhouping Wang, Jianrong Li, Guonan Chen
ABSTRAKT
A facile and feasible magneto-controlled immunosensing platform was designed for sensitive electrochemical immunoassay of brevetoxin B (BTX-2) in seafood by using guanine-assembled graphene nanoribbons (GGNRs) as molecular tags on a home-made magnetic carbon paste electrode. Initially, monoclonal mouse anti-BTX-2 antibodies were covalently immobilized on the surface of magnetic beads, which were used as the immunosensing probes for the capture of BTX-2. The recognition elements were prepared by chemical modification of bovine serum albumin-BTX-2 conjugates (BTX-2-BSA) with the GGNRs. Based on a competitive-type immunoassay format, the formed magnetic immunocomplex was integrated on the electrode with an external magnet, followed by determination in pH 6.5 phosphate-buffered solution containing 2 μM Ru(bpy)(3)Cl(2). Under optimal conditions, the electrochemical signals decreased with the increasing BTX-2 concentrations in the sample. The dynamic concentration range spanned from 1.0 pg mL(-1) to 10 ng mL(-1) with a detection limit of 1.0 pg mL(-1) BTX-2. Inter- and intra-batch assay precisions were substantially improved by resorting to the GGNR manifold. The method featured unbiased identification of negative (blank) and positive samples. No significant differences at the 95% confidence level were encountered in the analysis of 12 spiked samples including S. constricta, M. senhousia and T. granosa between the electrochemical immunoassay and commercially available enzyme-linked immunosorbent assay (ELISA) for determination of BTX-2.