Merck
Angiotensin (5-8) modulates nociception at the rat periaqueductal gray via the NO-sGC pathway and an endogenous opioid.
Neuroscience (2012-12-12)
L M Guethe, A Pelegrini-da-Silva, K G Borelli, M A Juliano, G G Pelosi, J B Pesquero, C L M Silva, F M A Corrêa, F Murad, W A Prado, A R Martins
ABSTRAKT
Angiotensins (Angs) modulate blood pressure, hydro-electrolyte composition, and antinociception. Although Ang (5-8) has generally been considered to be inactive, we show here that Ang (5-8) was the smallest Ang to elicit dose-dependent responses and receptor-mediated antinociception in the rat ventrolateral periaqueductal gray matter (vlPAG). Ang (5-8) antinociception seems to be selective, because it did not alter blood pressure or act on vascular or intestinal smooth muscle cells. The non-selective Ang-receptor (Ang-R) antagonist saralasin blocked Ang (5-8) antinociception, but selective antagonists of Ang-R types I, II, IV, and Mas did not, suggesting that Ang (5-8) may act via an unknown receptor. Endopeptidase EP 24.11 and amastatin-sensitive aminopeptidase from the vlPAG catalyzed the synthesis (from Ang II or Ang III) and inactivation of Ang (5-8), respectively. Selective inhibitors of neuronal-nitric oxide (NO) synthase, soluble guanylyl cyclase (sGC) and a non-selective opioid receptor (opioid-R) inhibitor blocked Ang (5-8)-induced antinociception. In conclusion, Ang (5-8) is a new member of the Ang family that selectively and strongly modulates antinociception via NO-sGC and endogenous opioid in the vlPAG.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Angiotensin Converting Enzyme Inhibitor, ≥95% (TLC)
Informacje o cenach i dostępności nie są obecnie dostępne.