Merck
Effect of three trifluoromethanesulfonate ionic liquids on the activity, stability and conformation of laccase.
International journal of biological macromolecules (2013-02-14)
Xinxin Yu, Feixue Zou, Ying Li, Lu Lu, Xirong Huang, Yinbo Qu
ABSTRAKT
The activity, stability and conformation of laccase were first investigated in an aqueous solution of ionic liquids 1-butyl-3-methylimidazolium trifluoromethanesulfonate ([Bmim]TfO), 1-butyl-1-methylpyrrolidinium trifluoromethanesulfonate ([Bmpyr]TfO) or tetramethylammonium trifluoromethanesulfonate ([TMA]TfO). Compared with control system, high level of [Bmim]TfO or [Bmpyr]TfO destabilizes laccase while [TMA]TfO stabilizes laccase. These effects are more pronounced with the extension of the incubation time. The activity variations are well correlated with the changes of the conformation of laccase evidenced by fluorescence and circular dichroism spectra under specified conditions. The effects of the three ionic liquids on laccase are associated with the chaotropicity of the cations in Hofmeister series. For laccase, [TMA]TfO is not a good activating agent but it greatly enhances the stability of laccase in addition to maintaining the catalytic efficiency of laccase, showing its great potential in real application.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Barium trifluoromethanesulfonate, 98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Laccase from Trametes versicolor, powder, light brown, ≥0.5 U/mg
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Laccase from Agaricus bisporus, powder, deep brown, ≥4 U/mg
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)