Merck
  • Sustained clinical resolution of acquired epidermodysplasia verruciformis in an immunocompromised patient after discontinuation of oral acitretin with topical imiquimod.
Sustained clinical resolution of acquired epidermodysplasia verruciformis in an immunocompromised patient after discontinuation of oral acitretin with topical imiquimod.
Journal of drugs in dermatology : JDD (2013-04-03)
Rajiv I Nijhawan, Achiamah Osei-Tutu, Jeremy M Hugh
ABSTRAKT
Increased cases of acquired epidermodysplasia verruciformis (EDV) have been reported in patients with human immunodeficiency virus (HIV). With regard to management, there are no randomized controlled trials in either immunocompetent or immunocompromised patients, and only a limited number of anecdotal treatment options. Systemic retinoids, either independently or in combination with other treatment modalities, have been used with limited success, demonstrating transient clinical response and recurrence of lesions after cessation of therapy. We report a case of an HIV-positive patient with acquired EDV who achieved sustained clinical resolution even after discontinuation of oral acitretin by applying topical imiquimod to prevent recurrence of his lesions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Imiquimod, ≥98% (HPLC), solid
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Acitretin, European Pharmacopoeia (EP) Reference Standard
USP
Acitretin Related Compound A, United States Pharmacopeia (USP) Reference Standard