Merck
Calcium binding cyclic hexapeptide. Crystal structure of cyclo-(L-prolyl-glycyl)3 calcium complex.
International journal of peptide and protein research (1986-02-01)
K I Varughese, S Aimoto, G Kartha
ABSTRAKT
The synthetic cyclic hexapeptide (L-prolyl-glycyl)3 forms a 2:1 complex with Ca2+ ion. The cation is sandwiched between the two peptide molecules. The glycyl carbonyls from each of the peptides are octahedrally coordinated to the cation with an average calcium oxygen coordination distance of 2.26A. Both the molecules coordinating to the calcium ion have three fold symmetry, but show significant conformational differences. In one of the peptides of the sandwich, the alternate carbonyls point to the opposite sides of the peptide ring while in the other, all the six carbonyls point to the same side of the ring. Three NH ... O hydrogen bonds between the peptides add to the stability of the sandwich.