Merck
Transport characteristics of p-acetamidobenzoic acid in brush border membrane of rat small intestine.
Journal of pharmacobio-dynamics (1983-11-01)
M Yasuhara, T Kimura, H Sezaki
ABSTRAKT
Brush border membrane vesicles of rat small intestinal mucosa were prepared by the modified calcium precipitation method in order to investigate the intestinal secretion mechanism of p-acetamidobenzoic acid (Ac-PABA). The release rate of [14C]-Ac-PABA from preloaded vesicles was much faster than that of p-aminobenzoic acid (PABA) and was enhanced by the addition of unlabelled Ac-PABA in the outer medium. In contrast, the membrane transfer rate of Ac-PABA across the egg lecithin liposomes was slower than that of PABA. The contribution of a carrier-mediated transport mechanism in the brush border membrane to the intestinal secretion of Ac-PABA was suggested.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%
Informacje o cenach i dostępności nie są obecnie dostępne.