Merck
Metabonomics approach to determine metabolic differences between green tea and black tea consumption.
Journal of agricultural and food chemistry (2006-08-31)
Ferdi A Van Dorsten, Clare A Daykin, Theo P J Mulder, John P M Van Duynhoven
ABSTRAKT
The purpose of this study was to compare the effects of black and green tea consumption on human metabolism. Seventeen healthy male volunteers consumed black tea, green tea, or caffeine in a randomized crossover study. Twenty-four-hour urine and blood plasma samples were analyzed by NMR-based metabonomics, that is, high-resolution 1H NMR metabolic profiling combined with multivariate statistics. Green and black tea consumption resulted in similar increases in urinary excretion of hippuric acid and 1,3-dihydroxyphenyl-2-O-sulfate, both of which are end products of tea flavonoid degradation by colonic bacteria. Several unidentified aromatic metabolites were detected in urine specifically after green tea intake. Interestingly, green and black tea intake also had a different impact on endogenous metabolites in urine and plasma. Green tea intake caused a stronger increase in urinary excretion of several citric acid cycle intermediates, which suggests an effect of green tea flavanols on human oxidative energy metabolism and/or biosynthetic pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Oxaloacetic acid, powder, suitable for hybridoma
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
SAFC
L-Tyrosine
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Tyrosine, FG
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.