Merck
Conjugation of benzoic acid with glycine in human liver and kidney: a study on the interindividual variability.
Xenobiotica; the fate of foreign compounds in biological systems (1993-12-01)
A Temellini, S Mogavero, P C Giulianotti, A Pietrabissa, F Mosca, G M Pacifici
ABSTRAKT
1. The rate of conjugation of benzoic acid with glycine was measured in the homogenates of 110 specimens of human liver and in 67 specimens of human renal cortex. 2. The assay for the formation of benzoyl glycine consisted of measuring the formation of benzoyl glycine from (14C) benzoic acid and glycine in the presence of coenzyme A and ATP. 3. In human liver, the mean (+/- SD) and coefficient of variation for the formation rate of benzoyl glycine were 254 +/- 90.5 nmol min-1 per g liver and 36%, respectively. There was a weak, but significant, negative correlation (r = -0.339, p < 0.001) between the rate of formation of benzoyl glycine and the liver donor's age. 4. In the human kidney, the rate of benzoyl glycine formation was normally distributed. The mean (+/- SD) and coefficient of variation were 321 +/- 99.3 nmol min-1 per g kidney and 31%, respectively. 5. These in vitro results are consistent with the view that the in vivo rate of conjugation of carboxylic acid with glycine varies among subjects and is normally distributed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%