Merck
Synthesis, spectral characterization and biological activity of metal(II) complexes with 4-aminoantipyrine derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-08)
C Leelavathy, S Arul Antony
ABSTRAKT
Novel metal(II) complexes derived from furfurylidene-4-aminoantipyrine and 2-aminobenzothiazole were synthesized and characterized by spectroscopic (IR, (1)H NMR, UV-Vis., ESR and DART-MS) and other analytical methods. IR spectral studies indicate the binding sites of the ligand with the metal ion. Molar conductance data and magnetic susceptibility measurements provide evidence for monomeric and neutral nature of the complexes. The X band ESR spectrum of the Cu(II) complex at 300 and 77K was recorded. The electrochemical behaviour of the complexes in MeCN at 298 K was studied. Thermal studies of the ligand and its complexes show the presence of coordinated water in the complexes. The grain size of the complex was calculated by Scherrer formula using powder XRD. The surface morphology of the complexes was studied using SEM. The in vitro biological screening of the ligand and its complexes were tested against bacterial species S. aureus, E. coli, K. pneumoniae, P. vulgaris and P. aeruginosa and fungal species A. niger, R. stolonifer, A. flavus, R. bataicola and C. albicans. The DNA binding and cleavage activity of the ligand and its complexes were studied. Super oxide dismutase (SOD) activities of the ligand and its complexes have also been measured.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%