Merck
In(III)/PhCO2H binary acid catalyzed tandem [2 + 2] cycloaddition and Nazarov reaction between alkynes and acetals.
Organic letters (2013-08-21)
Lihui Zhu, Zhi-Guo Xi, Jian Lv, Sanzhong Luo
ABSTRAKT
A facile tandem [2 + 2] cycloaddition and Nazarov reaction has been developed. The combination of In(OTf)3 and benzoic acid was found to synergistically promote the coupling of alkynes and acetals to form 2,3-disubstituted indanones in excellent yield and diastereoselectivity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Barium trifluoromethanesulfonate, 98%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%