Merck
Algibacter wandonensis sp. nov., isolated from sediment around a brown algae (Undaria pinnatifida) reservoir.
International journal of systematic and evolutionary microbiology (2013-09-03)
Jung-Hoon Yoon, Sooyeon Park
ABSTRAKT
A Gram-stain-negative, non-flagellated, rod-shaped bacterial strain able to move by gliding, designated WS-MY22(T), was isolated from sediment around a brown algae reservoir located on Wando in South Korea. It grew optimally at 25 °C, at pH 7.0-8.0 and in the presence of 2.0 % (w/v) NaCl. Phylogenetic trees based on 16S rRNA gene sequences showed that strain WS-MY22(T) clustered coherently with the type strains of Algibacter lectus and Algibacter undariae. It exhibited sequence similarity of 99.4 and 98.9 % to the type strains of A. lectus and A. undariae, respectively, and of 95.1-96.6 % to those of the other species of the genus Algibacter. Strain WS-MY22(T) contained MK-6 as the predominant menaquinone and iso-C15 : 1 G and iso-C17 : 0 3-OH as the major fatty acids. The major polar lipids of strain WS-MY22(T) were phosphatidylethanolamine and two unidentified lipids. The DNA G+C content of strain WS-MY22(T) was 35.8 mol% and its DNA-DNA relatedness with A. lectus KCTC 12103(T) and A. undariae WS-MY9(T) was 31 and 19 %, respectively. The phylogenetic and genetic distinctiveness and differential phenotypic properties revealed that strain WS-MY22(T) is separate from other species of the genus Algibacter. On the basis of the data presented, strain WS-MY22(T) is considered to represent a novel species of the genus Algibacter, for which the name Algibacter wandonensis sp. nov. is proposed. The type strain is WS-MY22(T) ( = KCTC 32381(T) = CECT 8301(T)).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder