Merck
Identification, characterization, and crystal structure of an aldo-keto reductase (AKR2E4) from the silkworm Bombyx mori.
Archives of biochemistry and biophysics (2013-09-10)
Kohji Yamamoto, David K Wilson
ABSTRAKT
A new member of the aldo-keto reductase (AKR) superfamily with 3-dehydroecdysone reductase activity was found in the silkworm Bombyx mori upon induction by the insecticide diazinon. The amino acid sequence showed that this enzyme belongs to the AKR2 family, and the protein was assigned the systematic name AKR2E4. In this study, recombinant AKR2E4 was expressed, purified to near homogeneity, and kinetically characterized. Additionally, its ternary structure in complex with NADP(+) and citrate was refined at 1.3Å resolution to elucidate substrate binding and catalysis. The enzyme is a 33-kDa monomer and reduces dicarbonyl compounds such as isatin and 17α-hydroxy progesterone using NADPH as a cosubstrate. No NADH-dependent activity was detected. Robust activity toward the substrate inhibitor 3-dehydroecdysone was observed, which suggests that this enzyme plays a role in regulation of the important molting hormone ecdysone. This structure constitutes the first insect AKR structure determined. Bound NADPH is located at the center of the TIM- or (β/α)8-barrel, and residues involved in catalysis are conserved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Ecdysone, ≥90%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast