Merck
Association of erythrocyte membrane fatty acids with changes in glycemia and risk of type 2 diabetes.
The American journal of clinical nutrition (2013-10-25)
Yuvaraj Mahendran, Jyrki Ågren, Matti Uusitupa, Henna Cederberg, Jagadish Vangipurapu, Alena Stančáková, Ursula Schwab, Johanna Kuusisto, Markku Laakso
ABSTRAKT
The significance of erythrocyte membrane fatty acids (EMFAs) and their ratios to predict hyperglycemia and incident type 2 diabetes is unclear. We investigated EMFAs as predictors of the worsening of hyperglycemia and incident type 2 diabetes in a 5-y follow-up of a population-based study. We measured EMFAs in 1346 Finnish men aged 45-73 y at baseline [mean ± SD age: 55 ± 6 y; body mass index (in kg/m(2)): 26.5 ± 3.5]. Our prospective follow-up study included only men who were nondiabetic at baseline and who had data available at the 5-y follow-up visit (n = 735). Our study showed that, after adjustment for confounding factors, palmitoleic acid (16:1n-7; P = 2.8 × 10(-7)), dihomo-γ-linolenic acid (20:3n-6; P = 2.3 × 10(-4)), the ratio of 16:1n-7 to 16:0 (P = 1.6 × 10(-8)) as a marker of stearoyl coenzyme A desaturase 1 activity, and the ratio of 20:3n-6 to 18:2n-6 (P = 9.4 × 10(-7)) as a marker of Δ(6)-desaturase activity significantly predicted the worsening of hyperglycemia (glucose area under the curve in an oral-glucose-tolerance test). In contrast, linoleic acid (18:2n-6; P = 0.0015) and the ratio of 18:1n-7 to 16:1n-7 (P = 1.5 × 10(-9)) as a marker of elongase activity had opposite associations. Statistical significance persisted even after adjustment for baseline insulin sensitivity, insulin secretion, and glycemia. Palmitoleic acid (P = 0.010) and the ratio of 16:1n-7 to 16:0 (P = 0.004) nominally predicted incident type 2 diabetes, whereas linoleic acid had an opposite association (P = 0.004), and n-3 polyunsaturated fatty acids did not show any associations. EMFAs and their ratios are associated longitudinally with changes in glycemia and the risk type 2 diabetes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Linoleic acid, ≥98%
Supelco
Palmitoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture