Merck
Distal connectivity causes summation and division across mouse visual cortex.
Nature neuroscience (2013-11-19)
Tatsuo K Sato, Michael Häusser, Matteo Carandini
ABSTRAKT
Neurons in different locations across the cortex are connected through polysynaptic networks involving both excitation and inhibition. To probe the functional effect of such networks, we used optogenetic stimulation to trigger antidromic spikes in a local region of primary visual cortex (V1). This local activity had two effects at distal V1 locations: summation and division. The balance between the two depended on visual contrast, and a normalization model precisely captured these effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)