Merck
Comparison of tolerability and adverse events following treatment with two GnRH agonists in patients with advanced prostate cancer.
Urologic nursing (2013-12-21)
Neal D Shore, Paul Sieber, Leanne Schimke, Adam Perzin, Scott Olsen
ABSTRAKT
This multicenter, randomized, crossover, open-label study (ClinicalTrials.gov identifier: NCT01161563) assessed patients'and clinicians'perceptions of injection site tolerability and adverse events following the intramuscular injection of triptorelin pamoate or subcutaneous injection of leuprolide acetate in 107 male, patients with advanced prostate cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Gonadorelin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Leuprolide acetate salt, ≥98% (HPLC)
Sigma-Aldrich
[D-Trp6]-LH-RH, ≥97% (HPLC), powder
Leuprorelin, European Pharmacopoeia (EP) Reference Standard