Merck
Degradation of oxidized and glycoxidized collagen: role of collagen cross-linking.
Archives of biochemistry and biophysics (2013-12-24)
Kerstin Nowotny, Tilman Grune
ABSTRAKT
Skin aging is a multifactorial process leading to structural and physiological changes. Protein modifications are known to play here an important role. Since collagen is the most abundant protein in the extracellular matrix of the skin and due to its slow turnover rates it is a frequent target of modifications by reactive compounds. Using skin biopsies of young and old mice we demonstrated that advanced glycation end products (AGEs), such as argpyrimidine and pentosidine, accumulate in aged skin, whereas protein carbonylation is unchanged. To investigate whether this discrepancy in accumulation is the result of an increased formation or due to reduced degradation we used modified collagen type I in in vitro experiments and tested for proteolytic susceptibility. We were able to show that collagenase is able to degrade oxidized and AGE-modified collagen. However, if collagen is cross-linked heavily, collagenase is unable to degrade the modified collagen. Cross-linking of collagen is preferentially taking place in collagen fibers treated with glycoxidizing agents. In summary, the low presence of oxidized collagen in aged skin seems to be the result of a sufficient degradation by collagenases, whereas the reason of the accumulation of AGE-modified collagen is at least partially an insufficient degradation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered