Merck
In vitro release of model compounds with different hydrophilicity from poly(ester-anhydride) microspheres.
Acta poloniae pharmaceutica (2014-01-05)
Katarzyna Jaszcz
ABSTRAKT
Poly(ester-anhydride) microspheres were prepared using emulsion solvent evaporation technique from two copolymers obtained by polycondensation of sebacic acid (SBA) and oligo(3-allyloxy-1,2-propylene succinate) terminated with carboxyl groups (OSAGE). The SBA content in copolymers was 90 or 70 w/w %, respectively. The size of microspheres obtained was in the range of 2-4 microm (small microspheres) or 12-31 microm (large ones) and depended on stirring conditions used in emulsion formulation process, as well as on concentration of polymer solution used. Poly(ester-anhydride) microspheres were loaded with three model compounds (rhodamine B, p-nitroaniline and piroxicam) with different water solubility. The loading efficiency was dependent on kind of model compound, polymer composition and size of microparticles. Each of three model compounds also exhibited different release profiles. Rhodamine was released very quickly. The release of p-nitroaniline was more prolonged, but showed only little dependence on microsphere size and polymer composition, whereas the piroxicam-loaded microspheres exhibited the most interesting release profile, showing that release rate as well as transport mechanism can be adjusted by changing microsphere size and poly(ester-anhydride) compositions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
Allyl glycidyl ether, ≥99%
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Piroxicam for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Piroxicam, ≥98% (TLC)
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Supelco
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitroaniline, ≥99%
Piroxicam, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
Sebacic acid, 99%
Sigma-Aldrich
Sebacic acid, ≥95.0% (GC)
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland