Merck
  • Elucidation of n-butyl benzyl phthalate biodegradation using high-performance liquid chromatography and gas chromatography-mass spectrometry.
Elucidation of n-butyl benzyl phthalate biodegradation using high-performance liquid chromatography and gas chromatography-mass spectrometry.
Analytical and bioanalytical chemistry (2006-07-28)
Xiang-Rong Xu, Hua-Bin Li, Ji-Dong Gu
ABSTRAKT
n-Butyl benzyl phthalate (BBP) is an endocrine-disrupting chemical. A bacterium species capable of using BBP as the sole source of carbon and energy was isolated from mangrove sediment. Effects of BBP concentration, pH, temperature, and salinity on BBP biodegradation were studied. The optimum pH, temperature, and salinity for the BBP biodegradation were 7.0, 37 degrees C, and 15 g L(-1), respectively. BBP was completely degraded within 6 days under optimum conditions, and the biodegradation of BBP could be fitted to a first-order kinetic model. The major metabolites of BBP biodegradation were identified as mono-butyl phthalate, mono-benzyl phthalate, phthalic acid, and benzoic acid by using high-performance liquid chromatography and gas chromatography-mass spectrometry. A preliminary metabolic pathway was proposed for the biodegradation of BBP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
mono-Benzyl phthalate, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.