Merck
  • Functional analysis of PsG6PDH, a cytosolic glucose-6-phosphate dehydrogenase gene from Populus suaveolens, and its contribution to cold tolerance improvement in tobacco plants.
Functional analysis of PsG6PDH, a cytosolic glucose-6-phosphate dehydrogenase gene from Populus suaveolens, and its contribution to cold tolerance improvement in tobacco plants.
Biotechnology letters (2013-05-22)
Yuanzhen Lin, Shanzhi Lin, Hai Guo, Zhiyi Zhang, Xiaoyang Chen
ABSTRAKT
A 1,697-bp cDNA sequence, designated as PsG6PDH, was amplified from Populus suaveolens. Multiple sequence alignment and phylogenetic analysis indicated that PsG6PDH encodes a cytosolic G6PDH isoform, with Southern blot analysis demonstrating that the gene is single or low copy in Populus. Transgenic tobacco plants over-expressing PsG6PDH exhibited enhanced cold tolerance. In both transgenic and wild-type (WT) tobacco plants, cold stress increased leaf malondialdehyde (MDA) content, electrolyte leakage (EL), and peroxide (POD) and superoxide dismutase (SOD) activities; relative to WT, however, transgenic lines had lower MDA content and EL and higher SOD and POD activities. In addition, PsG6PDH activated the expression of stress-related genes, including NtERD10b, NtERD10c, and NtSOD, in tobacco plants. Our results provide evidence regarding PsG6PDH regulatory function in plants during low temperature stress.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret