Merck
A real-time killing assay to follow viral epitope presentation to CD8 T cells.
Journal of immunological methods (2013-09-26)
Pauline Gourdain, Julie Boucau, Georgio Kourjian, Nicole Y Lai, Ellen Duong, Sylvie Le Gall
ABSTRAKT
The ability of cytotoxic T lymphocytes (CTL) to clear virus-infected cells requires the presentation of viral peptides intracellularly processed and displayed by major histocompatibility complex class I. Assays to measure CTL-mediated killing often use peptides exogenously added onto target cells--which does not account for epitope processing--or follow killing of infected cells at a single time point. In this study we established a real-time fluorogenic cytotoxic assay that measures the release of the Glucose-6-phosphate-dehydrogenase by dying target cells every 5 min after addition of CTL. It has comparable sensitivity to (51)chromium-based killing assay with the additional advantage of incorporating the kinetics of epitope presentation. We showed that HIV infection of immortalized or primary CD4 T cells leads to asynchronous killing by two CTL clones specific for epitopes located in different proteins. Real-time monitoring of killing of virus-infected cells will enable identification of immune responses efficiently preventing virus dissemination.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein