Merck
  • Surface-enhanced Raman spectroscopy-active substrates: adapting the shape of plasmonic nanoparticles for different biological applications.
Surface-enhanced Raman spectroscopy-active substrates: adapting the shape of plasmonic nanoparticles for different biological applications.
Journal of nanoscience and nanotechnology (2014-04-17)
Elina A Vitol, Gary Friedman, Yury Gogotsi
ABSTRAKT
We discuss the relationship between the shape of plasmonic nanoparticles and the biological surface-enhanced Raman spectroscopy (SERS) applications which they can enable. As a step forward in developing SERS-active substrates adapted to a particular application, we demonstrate that a modification of the widely used protocol for the sodium citrate mediated reduction of chloroauric acid, which is typically employed only for obtaining spherical gold nanoparticles, can yield flat polygonal nanoparticles at room temperature and a decreased amount of the reducing agent. The significant advantage of the described approach is that it allows for synthesis of nanoparticles with different geometries using a well-established synthesis protocol without the need for any additional chemicals or special synthesis apparatus. By contrasting spherical and anisotropically shaped nanoparticles, we demonstrate that multifaceted nanoparticles with sharp edges are better suitable for SERS analysis of low concentration analytes requiring strong SERS enhancement. On the other hand, gold nanoparticles with isotropic shapes, while giving a smaller enhancement, can provide a more reproducible SERS signal. This is important for analytical applications of complex biological systems where large SERS enhancement may not always be required, whereas data reproducibility and minimal false positive rate are imperative. Using a SERS-active substrate comprising isotropically shaped gold nanoparticles, we demonstrate the differences between Gram-negative (E. coli) and Gram-positive (S. aureus) bacteria, attributable to the outer membrane and peptidoglycan layer, with the level of detail which has not been previously reported with optical spectroscopic techniques.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)